Assay Details
| Gene Symbol: gna11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045036.2 | NP_001038501.1 | 7 - 8 | 1013 | 147 | |
| XM_005170474.4 | XP_005170531.1 | 6 - 7 | 966 | 147 | ||
| XM_005170475.4 | XP_005170532.1 | 6 - 7 | 890 | 147 | ||
| XM_021469340.1 | XP_021325015.1 | 5 - 6 | 638 | 147 | ||
More Information
Set Membership: |
|

gna11a
Genomic Map