Assay Details
| Gene Symbol: bik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045038.2 | NP_001038503.2 | 3 - 4 | 177 | 65 | |
| XM_005164756.1 | XP_005164813.1 | 3 - 4 | 267 | 65 | ||
| XM_009300298.1 | XP_009298573.1 | 3 - 4 | 595 | 65 | ||
| GenBank mRNA | BC135049.1 | - | 3 - 4 | 164 | 65 | |
| DQ860154.1 | - | 3 - 4 | 177 | 65 | ||
More Information
Set Membership: |
|

bik
Genomic Map