Assay Details
| Gene Symbol: bik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045038.2 | NP_001038503.2 | 4 - 5 | 325 | 112 | |
| XM_009300298.1 | XP_009298573.1 | 4 - 5 | 743 | 112 | ||
| GenBank mRNA | BC135049.1 | - | 4 - 5 | 312 | 112 | |
| DQ860154.1 | - | 4 - 5 | 325 | 112 | ||
More Information
Set Membership: |
|

bik
Genomic Map