Assay Details
| Gene Symbol: tulp4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045095.1 | NP_001038560.1 | 9 - 10 | 1595 | 128 | |
| XM_005168512.4 | XP_005168569.1 | 11 - 12 | 2295 | 128 | ||
More Information
Set Membership: |
|

tulp4a
Genomic Map