Assay Details
| Gene Symbol: tspan33b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045135.1 | NP_001038600.1 | 3 - 4 | 250 | 94 | |
| XM_005159028.4 | XP_005159085.1 | 3 - 4 | 426 | 94 | ||
More Information
Set Membership: |
|

tspan33b
Genomic Map