Assay Details
| Gene Symbol: scn8ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045183.1 | NP_001038648.1 | 6 - 7 | 932 | 68 | |
| XM_005166071.3 | XP_005166128.1 | 7 - 8 | 1206 | 68 | ||
| XM_005166072.3 | XP_005166129.1 | 7 - 8 | 1206 | 68 | ||
| XM_009302566.2 | XP_009300841.1 | 7 - 8 | 1205 | 68 | ||
| XM_021476941.1 | XP_021332616.1 | 7 - 8 | 1207 | 68 | ||
| XM_021476942.1 | XP_021332617.1 | 7 - 8 | 2593 | 68 | ||
| GenBank mRNA | DQ149509.1 | - | 6 - 7 | 932 | 68 | |
More Information
Set Membership: |
|

scn8ab
Genomic Map