Assay Details
| Gene Symbol: scn8ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045183.1 | NP_001038648.1 | 7 - 8 | 999 | 74 | |
| GenBank mRNA | DQ149509.1 | - | 7 - 8 | 999 | 74 | |
More Information
Set Membership: |
|

scn8ab
Genomic Map