Assay Details
| Gene Symbol: scn8ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045183.1 | NP_001038648.1 | 8 - 9 | 1138 | 108 | |
| XM_005166071.3 | XP_005166128.1 | 9 - 10 | 1412 | 108 | ||
| XM_005166072.3 | XP_005166129.1 | 9 - 10 | 1412 | 108 | ||
| XM_009302566.2 | XP_009300841.1 | 9 - 10 | 1411 | 108 | ||
| XM_021476941.1 | XP_021332616.1 | 9 - 10 | 1413 | 108 | ||
| XM_021476942.1 | XP_021332617.1 | 9 - 10 | 2799 | 108 | ||
| GenBank mRNA | DQ149509.1 | - | 8 - 9 | 1138 | 108 | |
More Information
Set Membership: |
|

scn8ab
Genomic Map