Assay Details
| Gene Symbol: ensaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045184.1 | NP_001038649.1 | 2 - 3 | 204 | 75 | |
| XM_005159315.4 | XP_005159372.1 | 2 - 3 | 324 | 75 | ||
| GenBank mRNA | BC162796.1 | - | 2 - 3 | 208 | 75 | |
More Information
Set Membership: |
|

ensaa
Genomic Map