Assay Details
| Gene Symbol: nrm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045195.1 | NP_001038660.1 | 3 - 4 | 515 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nrm| Gene Symbol: nrm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045195.1 | NP_001038660.1 | 3 - 4 | 515 | 74 | |
Set Membership: |
|