Assay Details
| Gene Symbol: fam102ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045316.1 | NP_001038781.1 | 9 - 10 | 1096 | 74 | |
| GenBank mRNA | BC117658.1 | - | 9 - 10 | 1096 | 74 | |
More Information
Set Membership: |
|

fam102ba
Genomic Map