Assay Details
| Gene Symbol: b4galt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045332.1 | NP_001038797.1 | 3 - 4 | 589 | 64 | |
| GenBank mRNA | BC163615.1 | - | 3 - 4 | 500 | 64 | |
| DQ104219.1 | - | 3 - 4 | 589 | 64 | ||
More Information
Set Membership: |
|

b4galt5
Genomic Map