Assay Details
| Gene Symbol: mrps9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045396.1 | NP_001038861.1 | 5 - 6 | 511 | 67 | |
| GenBank mRNA | BC122171.1 | - | 5 - 6 | 511 | 67 | |
| BC165785.1 | - | 5 - 6 | 488 | 67 | ||
More Information
Set Membership: |
|

mrps9
Genomic Map