Assay Details
| Gene Symbol: tpk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045397.1 | NP_001038862.1 | 2 - 3 | 435 | 86 | |
| GenBank mRNA | BC122452.1 | - | 2 - 3 | 435 | 86 | |
More Information
Set Membership: |
|

tpk2
Genomic Map