Assay Details
| Gene Symbol: tpk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045397.1 | NP_001038862.1 | 6 - 7 | 883 | 74 | |
| GenBank mRNA | BC092735.1 | - | 6 - 7 | 587 | 74 | |
| BC122452.1 | - | 6 - 7 | 883 | 74 | ||
More Information
Set Membership: |
|

tpk2
Genomic Map