Assay Details
| Gene Symbol: mybpc2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045859.1 | NP_001039324.1 | 22 - 23 | 2449 | 90 | |
| XM_021471480.1 | XP_021327155.1 | 24 - 25 | 2553 | 90 | ||
| GenBank mRNA | BC121195.1 | - | 22 - 23 | 2449 | 90 | |
More Information
Set Membership: |
|

mybpc2a
Genomic Map