Assay Details
| Gene Symbol: galnt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076562.2 | NP_001070030.2 | 8 - 9 | 1306 | 57 | |
| GenBank mRNA | BC124297.1 | - | 8 - 9 | 1303 | 57 | |
| EH485353.1 | - | 5 - 6 | 722 | 57 | ||
More Information
Set Membership: |
|

galnt11
Genomic Map