Assay Details
| Gene Symbol: sgk2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076564.1 | NP_001070032.1 | 5 - 6 | 315 | 86 | |
| XM_009292340.3 | XP_009290615.1 | 5 - 6 | 384 | 86 | ||
| GenBank mRNA | BC124529.1 | - | 5 - 6 | 315 | 86 | |
| BC164500.1 | - | 5 - 6 | 299 | 86 | ||
More Information
Set Membership: |
|

sgk2b
Genomic Map