Assay Details
| Gene Symbol: ccnyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076720.1 | NP_001070188.1 | 7 - 8 | 667 | 61 | |
| GenBank mRNA | BC124226.1 | - | 7 - 8 | 667 | 61 | |
More Information
Set Membership: |
|

ccnyl1
Genomic Map