Assay Details
Gene Symbol: kaznb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080082.2 | NP_001073551.2 | 3 - 4 | 839 | 73 | |
XM_009306042.3 | XP_009304317.1 | 2 - 3 | 1070 | 73 | ||
XM_009306043.3 | XP_009304318.2 | 6 - 7 | 997 | 73 | ||
XM_017358249.2 | XP_017213738.1 | 6 - 7 | 1008 | 73 | ||
XM_017358250.2 | XP_017213739.1 | 6 - 7 | 997 | 73 | ||
XM_021479663.1 | XP_021335338.1 | 3 - 4 | 509 | 73 | ||
GenBank mRNA | BC128836.1 | - | 3 - 4 | 674 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
