Assay Details
Gene Symbol: kaznb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080082.2 | NP_001073551.2 | 7 - 8 | 1461 | 100 | |
XM_009306042.3 | XP_009304317.1 | 6 - 7 | 1692 | 100 | ||
XM_017358249.2 | XP_017213738.1 | 10 - 11 | 1630 | 100 | ||
XM_017358250.2 | XP_017213739.1 | 10 - 11 | 1619 | 100 | ||
XM_021479663.1 | XP_021335338.1 | 7 - 8 | 1131 | 100 | ||
GenBank mRNA | BC128836.1 | - | 7 - 8 | 1296 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
