Assay Details
| Gene Symbol: slc6a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080580.1 | NP_001074049.1 | 5 - 6 | 757 | 64 | |
| XM_005161302.4 | XP_005161359.1 | 5 - 6 | 1082 | 64 | ||
| GenBank mRNA | BC129163.1 | - | 5 - 6 | 757 | 64 | |
More Information
Set Membership: |
|

slc6a7
Genomic Map