Assay Details
| Gene Symbol: fzd3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080601.1 | NP_001074070.1 | 4 - 5 | 1592 | 81 | |
| XM_005158594.4 | XP_005158651.1 | 5 - 6 | 2273 | 81 | ||
| GenBank mRNA | AB246777.1 | - | 4 - 5 | 1592 | 81 | |
| BC154488.1 | - | 5 - 6 | 2196 | 81 | ||
| BC163423.1 | - | 4 - 5 | 1663 | 81 | ||
| BC163436.1 | - | 4 - 5 | 1663 | 81 | ||
More Information
Set Membership: |
|

fzd3b
Genomic Map