Assay Details
| Gene Symbol: pank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080606.2 | NP_001074075.2 | 6 - 7 | 1425 | 81 | |
| XM_005156572.3 | XP_005156629.1 | 6 - 7 | 1149 | 81 | ||
| XM_005156573.3 | XP_005156630.1 | 5 - 6 | 815 | 81 | ||
| GenBank mRNA | BC129366.1 | - | 6 - 7 | 1418 | 81 | |
More Information
Set Membership: |
|

pank2
Genomic Map