Assay Details
| Gene Symbol: napepld | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080613.2 | NP_001074082.2 | 2 - 3 | 303 | 83 | |
| XM_005164895.4 | XP_005164952.1 | 3 - 4 | 628 | 83 | ||
| XM_005164897.4 | XP_005164954.1 | 3 - 4 | 504 | 83 | ||
| XM_021475123.1 | XP_021330798.1 | 3 - 4 | 485 | 83 | ||
| GenBank mRNA | BC129493.1 | - | 2 - 3 | 386 | 83 | |
| BC164764.1 | - | 1 - 2 | 237 | 83 | ||
| EB868982.1 | - | 2 - 3 | 281 | 83 | ||
More Information
Set Membership: |
|

napepld
Genomic Map