Assay Details
| Gene Symbol: ppat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082877.2 | NP_001076346.2 | 3 - 4 | 759 | 75 | |
| GenBank mRNA | BC142820.1 | - | 3 - 4 | 432 | 75 | |
| BC146730.1 | - | 3 - 4 | 759 | 75 | ||
More Information
Set Membership: |
|

ppat
Genomic Map