Assay Details
| Gene Symbol: bud23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082879.1 | NP_001076348.1 | 4 - 5 | 377 | 103 | |
| GenBank mRNA | BC133941.1 | - | 4 - 5 | 377 | 103 | |
More Information
Set Membership: |
|

bud23
Genomic Map