Assay Details
| Gene Symbol: fosl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082998.2 | NP_001076467.1 | 1 - 2 | 592 | 115 | |
| NM_001362607.1 | NP_001349536.1 | 2 - 3 | 249 | 115 | ||
| GenBank mRNA | BC133086.1 | - | 1 - 2 | 580 | 115 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001362607.1 transcripts.
Set Membership: |
|

fosl2
Genomic Map