Assay Details
| Gene Symbol: dpagt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089411.1 | NP_001082880.1 | 6 - 7 | 1236 | 61 | |
| GenBank mRNA | BC139558.1 | - | 6 - 7 | 1236 | 61 | |
More Information
Set Membership: |
|

dpagt1
Genomic Map