Assay Details
| Gene Symbol: myo1ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100031.1 | NP_001093501.1 | 10 - 11 | 1284 | 98 | |
| XM_005165347.4 | XP_005165404.1 | 10 - 11 | 1453 | 98 | ||
| XM_005165349.4 | XP_005165406.1 | 10 - 11 | 1513 | 98 | ||
More Information
Set Membership: |
|

myo1ca
Genomic Map