Assay Details
| Gene Symbol: myo1ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100031.1 | NP_001093501.1 | 16 - 17 | 1781 | 104 | |
| XM_005165347.4 | XP_005165404.1 | 16 - 17 | 1956 | 104 | ||
| XM_005165349.4 | XP_005165406.1 | 16 - 17 | 2016 | 104 | ||
More Information
Set Membership: |
|

myo1ca
Genomic Map