Assay Details
| Gene Symbol: paqr7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100060.1 | NP_001093530.1 | 1 | 957 | 65 | |
| XM_005173773.3 | XP_005173830.1 | 2 | 1121 | 65 | ||
More Information
Set Membership: |
|

paqr7a
Genomic Map