Assay Details
| Gene Symbol: mylka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009304445.1 | XP_009302720.1 | 22 - 23 | 4205 | 75 | |
| XM_009304446.1 | XP_009302721.1 | 22 - 23 | 4205 | 75 | ||
| XM_017357739.1 | XP_017213228.1 | 22 - 23 | 4202 | 75 | ||
| GenBank mRNA | AB267908.1 | - | 7 - 8 | 1562 | 75 | |
More Information
Set Membership: |
|

mylka
Genomic Map