Assay Details
| Gene Symbol: nagk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105693.1 | NP_001099163.1 | 9 - 10 | 874 | 91 | |
| GenBank mRNA | BC151923.1 | - | 9 - 10 | 874 | 91 | |
More Information
Set Membership: |
|

nagk
Genomic Map