Assay Details
| Gene Symbol: nagpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109715.2 | NP_001103185.2 | 6 - 7 | 1204 | 68 | |
| GenBank mRNA | BC152498.1 | - | 6 - 7 | 1197 | 68 | |
More Information
Set Membership: |
|

nagpa
Genomic Map