Assay Details
| Gene Symbol: wscd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109769.1 | NP_001103239.1 | 3 - 4 | 705 | 67 | |
| XM_005165158.4 | XP_005165215.1 | 6 - 7 | 1812 | 67 | ||
More Information
Set Membership: |
|

wscd2
Genomic Map