Assay Details
| Gene Symbol: lhfpl5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110113.2 | NP_001103583.2 | 1 - 2 | 504 | 82 | |
| GenBank mRNA | BC153982.1 | - | 1 - 2 | 504 | 82 | |
More Information
Set Membership: |
|

lhfpl5b
Genomic Map