Assay Details
| Gene Symbol: en2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131040.1 | NP_571115.1 | 1 - 2 | 807 | 101 | |
| GenBank mRNA | BC071290.1 | - | 1 - 2 | 819 | 101 | |
| X68447.1 | - | 807 | 101 | |||
More Information
Set Membership: |
|

en2b
Genomic Map