Assay Details
| Gene Symbol: nr2f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131183.1 | NP_571258.1 | 2 - 3 | 1332 | 86 | |
| GenBank mRNA | BC162484.1 | - | 2 - 3 | 1183 | 86 | |
| S80986.1 | - | 2 - 3 | 1332 | 86 | ||
More Information
Set Membership: |
|

nr2f2
Genomic Map