Assay Details
| Gene Symbol: csnk2a2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131240.1 | NP_571315.1 | 11 - 12 | 1942 | 84 | |
| GenBank mRNA | BC044342.1 | - | 11 - 12 | 1925 | 84 | |
| X99964.1 | - | 1942 | 84 | |||
More Information
Set Membership: |
|

csnk2a2a
Genomic Map