Assay Details
| Gene Symbol: csnk2a2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131240.1 | NP_571315.1 | 9 - 10 | 1708 | 92 | |
| GenBank mRNA | BC044342.1 | - | 9 - 10 | 1691 | 92 | |
| BC164160.1 | - | 9 - 10 | 858 | 92 | ||
| X99964.1 | - | 1708 | 92 | |||
More Information
Set Membership: |
|

csnk2a2a
Genomic Map