Assay Details
| Gene Symbol: rack1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131444.1 | NP_571519.1 | 6 - 7 | 837 | 67 | |
| GenBank mRNA | AF025330.1 | - | 6 - 7 | 837 | 67 | |
| AY423038.1 | - | 6 - 7 | 844 | 67 | ||
| BC049459.1 | - | 6 - 7 | 824 | 67 | ||
| BC092910.1 | - | 1225 | 67 | |||
More Information
Set Membership: |
|

rack1
Genomic Map