Assay Details
| Gene Symbol: dazl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131524.1 | NP_571599.1 | 8 - 9 | 901 | 66 | |
| XM_005170070.4 | XP_005170127.1 | 8 - 9 | 850 | 66 | ||
| XM_005170071.3 | XP_005170128.1 | 8 - 9 | 696 | 66 | ||
| XM_005170072.4 | XP_005170129.1 | 8 - 9 | 692 | 66 | ||
| GenBank mRNA | AB018191.1 | - | 8 - 9 | 901 | 66 | |
| BC076423.1 | - | 8 - 9 | 776 | 66 | ||
More Information
Set Membership: |
|

dazl
Genomic Map