Assay Details
| Gene Symbol: mapk14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131722.1 | NP_571797.1 | 1 - 2 | 362 | 80 | |
| XM_005167061.4 | XP_005167118.1 | 1 - 2 | 493 | 80 | ||
| GenBank mRNA | AB030897.1 | - | 1 - 2 | 362 | 80 | |
| BC044128.1 | - | 1 - 2 | 80 | |||
| BC059417.1 | - | 1 - 2 | 366 | 80 | ||
| BC164402.1 | - | 1 - 2 | 80 | |||
More Information
Set Membership: |
|

mapk14a
Genomic Map