Assay Details
| Gene Symbol: mapk14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131722.1 | NP_571797.1 | 5 - 6 | 685 | 139 | |
| XM_005167061.4 | XP_005167118.1 | 5 - 6 | 816 | 139 | ||
| GenBank mRNA | AB030897.1 | - | 5 - 6 | 685 | 139 | |
| BC044128.1 | - | 5 - 6 | 520 | 139 | ||
| BC059417.1 | - | 5 - 6 | 689 | 139 | ||
| BC164402.1 | - | 5 - 6 | 479 | 139 | ||
More Information
Set Membership: |
|

mapk14a
Genomic Map