Assay Details
| Gene Symbol: mapk14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131722.1 | NP_571797.1 | 9 - 10 | 1007 | 98 | |
| GenBank mRNA | AB030897.1 | - | 9 - 10 | 1007 | 98 | |
| BC044128.1 | - | 9 - 10 | 842 | 98 | ||
| BC059417.1 | - | 9 - 10 | 1012 | 98 | ||
| BC164402.1 | - | 9 - 10 | 801 | 98 | ||
More Information
Set Membership: |
|

mapk14a
Genomic Map