Assay Details
| Gene Symbol: epha4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153658.1 | NP_705944.1 | 6 - 7 | 1468 | 90 | |
| GenBank mRNA | AF369381.1 | - | 6 - 7 | 1468 | 90 | |
| Gene Symbol: LOC101882343 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005174542.2 | XP_005174599.2 | 1 - 2 | 190 | 90 | |
More Information
Set Membership: |
|

epha4b
Genomic Map