Assay Details
| Gene Symbol: epha4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153658.1 | NP_705944.1 | 7 - 8 | 1632 | 87 | |
| GenBank mRNA | AF369381.1 | - | 7 - 8 | 1632 | 87 | |
More Information
Set Membership: |
|

epha4b
Genomic Map