Assay Details
| Gene Symbol: aldh3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173221.2 | NP_775328.2 | 4 - 5 | 508 | 124 | |
| XM_005163981.4 | XP_005164038.1 | 4 - 5 | 552 | 124 | ||
| GenBank mRNA | AF254955.1 | - | 3 - 4 | 402 | 124 | |
| BC049338.1 | - | 4 - 5 | 508 | 124 | ||
More Information
Set Membership: |
|

aldh3b1
Genomic Map