Assay Details
| Gene Symbol: aldh3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173221.2 | NP_775328.2 | 7 - 8 | 1069 | 140 | |
| XM_005163981.4 | XP_005164038.1 | 7 - 8 | 1113 | 140 | ||
| GenBank mRNA | AF254955.1 | - | 6 - 7 | 963 | 140 | |
| BC049338.1 | - | 7 - 8 | 1069 | 140 | ||
More Information
Set Membership: |
|

aldh3b1
Genomic Map